Methyl-5-Norbornene-2-carboxylate (endo and exo mixture) 99% (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: 5-Norbornene-2-carboxylic Acid Methyl Ester, Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid Methyl Ester, Methyl Bicyclo[2.2.1]hept-5-ene-2-carboxylate, Methyl bicyclo[2.2.1]hept-5-ene-2-carboxylate

A1476244-100G A1476244-1G A1476244-5G
76663-752EA 290.69 USD
76663-752 76663-754 76663-756
Methyl-5-Norbornene-2-carboxylate (endo and exo mixture) 99% (mixture of isomers)
Methyl-5-Norbornene-2-carboxylate (endo and exo mixture)

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00167592
CAS Number: 6203-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR