(R)-4-Benzyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1476159-100MG A1476159-1G A1476159-250MG
76663-694EA 142 USD
76663-694 76663-696 76663-698
(R)-4-Benzyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(R)-4-Benzyl-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O
MW: 252.31 g/mol
CAS Number: 2757082-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR