(R)-4-(tert-Butyl)-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole 97%
Supplier: Ambeed
|
Warning
|
A1476157-100MG
A1476157-1G
A1476157-250MG
76663-682EA
150.65
USD
76663-682
76663-684
76663-686
(R)-4-(tert-Butyl)-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole 97%
(R)-4-(tert-Butyl)-2-(6-methylpyridin-2-yl)-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₁₃H₁₈N₂O MW: 218.2948 g/mol |
CAS Number:
2491654-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...