Methyl 6-bromo-2-fluoronicotinate 95%

Supplier: Ambeed
Danger

A1476133-100MG A1476133-5G A1476133-1G A1476133-250MG
76663-670EA 28 USD
76663-670 77567-048 77592-772 77875-201
Methyl 6-bromo-2-fluoronicotinate 95%
Methyl 6-bromo-2-fluoronicotinate

Precaución: For research use only.

Formula: C₇H₅BrFNO₂
MDL Number: MFCD14698122
CAS Number: 1214385-82-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR