[4,4'-Bis(trifluoromethyl)-2,2'-bipyridine-κN1,κN1']bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]Iridium hexafluorophosphate 97%
Supplier: Ambeed
|
Warning
|
A1476095-100MG
A1476095-1G
A1476095-250MG
A1476095-50MG
76663-662EA
160
USD
76663-662
76663-664
76663-666
76663-668
[4,4'-Bis(trifluoromethyl)-2,2'-bipyridine-κN1,κN1']bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]Iridium hexafluorophosphate 97%
[4,4'-Bis(trifluoromethyl)-2,2'-bipyridine-κN1,κN1']bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]Iridium hexafluorophosphate
Precaución: for Research Use Only
Formula:
C₃₄H₁₆F₁₈IrN₄P MW: 1045.6797 g/mol |
CAS Number:
2229858-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...