6,7-Dimethoxyquinolin-4(1H)-one 98%

Supplier: Ambeed
Warning

A1476073-1G A1476073-5G
76663-644EA 16.98 USD
76663-644 76663-646
6,7-Dimethoxyquinolin-4(1H)-one 98%
6,7-Dimethoxyquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
MDL Number: MFCD09031937
CAS Number: 127285-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR