1H-Indazole-6-carboxylic acid 98+%

Supplier: Ambeed
Warning

A1476065-100G A1476065-25G A1476065-5G
76663-632EA 575.01 USD
76663-632 76663-634 76663-636
1H-Indazole-6-carboxylic acid 98+%
1H-Indazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.1454 g/mol
MDL Number: MFCD06804571
CAS Number: 704-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR