7-Amino-1H-indazole 98+%

Supplier: Ambeed
Warning

A1476063-100G A1476063-25G A1476063-5G
76663-626EA 684.29 USD
76663-626 76663-628 76663-630
7-Amino-1H-indazole 98+%
7-Amino-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD00022790
CAS Number: 21443-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR