1-(4-Chlorobenzhydryl)piperazine 98% + (contains < 7% H₂O)
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Chlorodiphenylmethyl)piperazine, 1-((4-Chlorophenyl)(phenyl)methyl)piperazine
A1476049-1000G
A1476049-100G
A1476049-25G
A1476049-500G
76663-618EA
263.11
USD
76663-618
76663-620
76663-622
76663-624
1-(4-Chlorobenzhydryl)piperazine 98% + (contains < 7% H₂O)
1-(4-Chlorobenzhydryl)piperazine
Precaución: for Research Use Only
Formula:
C₁₇H₁₉ClN₂ MW: 286.80 g/mol |
MDL Number:
MFCD00005965 CAS Number: 303-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...