4-Bromo-1-methyl-1H-pyrazole 99%

Supplier: Ambeed
Warning

A1476043-100G A1476043-25G A1476043-500G A1476043-10G
76663-610EA 17 USD
76663-610 76663-612 76663-614 77624-324
4-Bromo-1-methyl-1H-pyrazole 99%
4-Bromo-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅BrN₂
MDL Number: MFCD02179565
CAS Number: 15803-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR