4-Methyl-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A1476033-100G A1476033-1G A1476033-25G A1476033-5G
76663-584EA 1194.58 USD
76663-584 76663-586 77597-494 77624-322
4-Methyl-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MDL Number: MFCD13619883
CAS Number: 945950-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR