2-Amino-5-bromobenzophenone 99+%

Supplier: Ambeed
Warning

A1476027-100G A1476027-25G A1476027-5G
76663-572EA 43 USD
76663-572 76663-574 76663-576
2-Amino-5-bromobenzophenone 99+%
2-Amino-5-bromobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrNO
MDL Number: MFCD00059962
CAS Number: 39859-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR