2-Bromo-1-[4-hydroxy-3-(hydroxymethyl) phenyl]ethan-1-one 95+%

Supplier: Ambeed
Danger

A1476023-25G A1476023-5G
76663-570EA 219.61 USD
76663-570 77592-770
2-Bromo-1-[4-hydroxy-3-(hydroxymethyl) phenyl]ethan-1-one 95+%
2-Bromo-1-[4-hydroxy-3-(hydroxymethyl) phenyl]ethan-1-one

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD18976157
CAS Number: 62932-94-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR