N-Carbethoxy-4-piperidone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Piperidone-N-carboxylic acid ethyl ester, N-Ethoxycarbonyl-4-piperidone, Ethyl4-oxo-1-piperidinecarboxylate

A1476015-100G A1476015-25G A1476015-500G
76663-556EA 19.1 USD
76663-556 76663-558 76663-560
N-Carbethoxy-4-piperidone 98%
N-Carbethoxy-4-piperidone

Precaución: for Research Use Only

Formula: C₈H₁₃NO₃
MW: 171.19 g/mol
MDL Number: MFCD00006188
CAS Number: 29976-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR