5-Chloro-2-methylanisole ≥99%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-methoxy-1-methylbenzene

A1475898-25G A1475898-5G
76663-538EA 194.15 USD
76663-538 76663-540
5-Chloro-2-methylanisole ≥99%
5-Chloro-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.6095 g/mol
MDL Number: MFCD07368289
CAS Number: 40794-04-5
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR