2-Bromo-4-(trifluoromethoxy)benzonitrile 99+%

Supplier: Ambeed
Warning

A1475895-100G A1475895-1G A1475895-25G A1475895-5G
76663-522EA 455 USD
76663-522 76663-524 76663-526 76663-528
2-Bromo-4-(trifluoromethoxy)benzonitrile 99+%
2-Bromo-4-(trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrF₃NO
MDL Number: MFCD13185874
CAS Number: 1214334-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR