Ethyl 3-fluoro-4-methylbenzoate 99+%

Supplier: Ambeed
Warning

A1475894-5G A1475894-25G
76663-520EA 47 USD
76663-520 77875-190
Ethyl 3-fluoro-4-methylbenzoate 99+%
Ethyl 3-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MDL Number: MFCD06204523
CAS Number: 86239-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR