Ethyl 4-bromo-3-fluorobenzoate 99%

Supplier: Ambeed
Warning

A1475890-100G A1475890-25G A1475890-5G
76663-500EA 363.89 USD
76663-500 76663-502 76663-504
Ethyl 4-bromo-3-fluorobenzoate 99%
Ethyl 4-bromo-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
MDL Number: MFCD09751983
CAS Number: 1130165-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR