3-(Difluoromethoxy)benzonitrile 99+%

Supplier: Ambeed
Warning

A1475888-100G A1475888-1G A1475888-25G A1475888-5G
76663-492EA 328 USD
76663-492 76663-494 76663-496 76663-498
3-(Difluoromethoxy)benzonitrile 99+%
3-(Difluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂NO
MDL Number: MFCD00236225
CAS Number: 97582-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR