2-Bromo-4-fluoro-5-methylbenzoic acid 99+%

Supplier: Ambeed
Warning

A1475881-100G A1475881-1G A1475881-25G A1475881-5G
76663-470EA 479 USD
76663-470 76663-472 76663-474 76663-476
2-Bromo-4-fluoro-5-methylbenzoic acid 99+%
2-Bromo-4-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MDL Number: MFCD09263434
CAS Number: 1003709-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR