2-Bromo-3,4,5-trifluoroaniline 99+%

Supplier: Ambeed
Warning

A1475867-100G A1475867-25G A1475867-5G
76663-446EA 2067.7 USD
76663-446 76663-448 76663-450
2-Bromo-3,4,5-trifluoroaniline 99+%
2-Bromo-3,4,5-trifluoroaniline

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MW: 225.9939 g/mol
MDL Number: MFCD01311010
CAS Number: 1000577-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR