4-Fluoro-m-toluonitrile 99%

Supplier: Ambeed
Warning

A1475862-100G A1475862-25G A1475862-5G
76663-430EA 190.97 USD
76663-430 76663-432 76663-434
4-Fluoro-m-toluonitrile 99%
4-Fluoro-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD03788510
CAS Number: 185147-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR