Ethyl 4-bromo-2-fluorobenzoate 99+%

Supplier: Ambeed
Warning

A1475841-100G A1475841-25G A1475841-5G
76663-398EA 119 USD
76663-398 76663-400 76663-402
Ethyl 4-bromo-2-fluorobenzoate 99+%
Ethyl 4-bromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MDL Number: MFCD06204444
CAS Number: 474709-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR