2-(Trifluoromethyl)phenylacetonitrile 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Trifluoromethyl)benzyl cyanide, 1-(Cyanmethyl)-2-(trifluormethyl)benzene
A1475835-100G
A1475835-25G
A1475835-5G
76663-392EA
291.75
USD
76663-392
76663-394
76663-396
2-(Trifluoromethyl)phenylacetonitrile 99+%
2-(Trifluoromethyl)phenylacetonitrile
Precaución: for Research Use Only
Formula:
C₉H₆F₃N MW: 185.15 g/mol |
MDL Number:
MFCD00001903 CAS Number: 3038-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...