Ethyl benzo[d]thiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

A1475811-100MG A1475811-1G A1475811-250MG A1475811-25G A1475811-5G
76663-366EA 21.22 USD
76663-366 76663-368 76663-370 76663-372 76663-374
Ethyl benzo[d]thiazole-5-carboxylate 98%
Ethyl benzo[d]thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂S
MW: 207.24 g/mol
MDL Number: MFCD13176733
CAS Number: 103261-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR