Aluminum monostearate ~70% (Al), Analytical reagent (AR)

Supplier: Ambeed
Warning

Synonyms: Dihydroxy(stearoyloxy)aluminum, Aluminum monostearate technical(Hard and soft aluminium mixture), Aluminum dihydroxide stearate

A1475751-100G A1475751-25G A1475751-500G
76663-360EA 22 USD
76663-360 76663-362 76663-364
Aluminum monostearate ~70% (Al), Analytical reagent (AR)
Aluminum monostearate

Precaución: for Research Use Only

Formula: C₁₈H₃₇AlO₄
MDL Number: MFCD00019932
CAS Number: 7047-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR