3-Fluoro-p-toluonitrile 98+%

Supplier: Ambeed
Danger

A1475729-25G A1475729-5G
76663-348EA 12 USD
76663-348 76663-350
3-Fluoro-p-toluonitrile 98+%
3-Fluoro-p-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
MDL Number: MFCD00153174
CAS Number: 170572-49-3
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR