2,3-Difluoro-4-ethoxybenzonitrile 98+%

Supplier: Ambeed
Warning

A1475725-100G A1475725-1G A1475725-25G A1475725-5G
76663-340EA 233 USD
76663-340 76663-342 76663-344 76663-346
2,3-Difluoro-4-ethoxybenzonitrile 98+%
2,3-Difluoro-4-ethoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₇F₂NO
MDL Number: MFCD04973770
CAS Number: 126162-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR