Acriflavine Cl,13.3-15.8%

Supplier: Ambeed
Danger

Synonyms: 3,6-Diamino-10-methylacridinium chloride, 3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mix, Acriflavinium chloride 3,6-Acridinediamine mix, 3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral, Euflavine, Acriflavine base

A1475625-25G A1475625-5G
76663-338EA 62.6 USD
76663-338 77597-472
Acriflavine Cl,13.3-15.8%
Acriflavine

Precaución: for Research Use Only

Formula: C₂₇H₂₅ClN₆
MW: 468.9806 g/mol
MDL Number: MFCD00064307
CAS Number: 8048-52-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR