2,4,6-Triisopropylthiophenol 97%

Supplier: Ambeed
Danger

Synonyms: 2,4,6-Triisopropylbenzenethiol

A1475513-1G A1475513-25G A1475513-5G
76663-324EA 28.65 USD
76663-324 76663-326 77549-812
2,4,6-Triisopropylthiophenol 97%
2,4,6-Triisopropylthiophenol

Precaución: for Research Use Only

Formula: C₁₅H₂₄S
MW: 236.4161 g/mol
MDL Number: MFCD12026227
CAS Number: 22693-41-0
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR