4-(Dibenzo[b,d]furan-4-yl)-N-(4-(naphthalen-1-yl)phenyl)aniline 98%

Supplier: Ambeed
Warning

A1475401-1G A1475401-5G
76663-320EA 5 USD
76663-320 76663-322
4-(Dibenzo[b,d]furan-4-yl)-N-(4-(naphthalen-1-yl)phenyl)aniline 98%
4-(Dibenzo[b,d]furan-4-yl)-N-(4-(naphthalen-1-yl)phenyl)aniline

Precaución: for Research Use Only

Formula: C₃₄H₂₃NO
MDL Number: MFCD32709943
CAS Number: 2097371-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR