2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride (contains 10-15% H₂O)

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride, 3-(2-Oxo-1,2-dihydro-4-quinolinyl)-DL-alanine hydrochloride

A1475356-100G A1475356-25G A1475356-5G
76663-314EA 62 USD
76663-314 76663-316 76663-318
2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride (contains 10-15% H₂O)
2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClN₂O₃
MW: 268.70 g/mol
MDL Number: MFCD07787426
CAS Number: 4876-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR