Methyl trans-3-Amino-cyclobutanecarboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A1475326-100MG A1475326-250MG A1475326-5G A1475326-1G
76663-308EA 9 USD
76663-308 76663-310 76663-312 77567-042
Methyl trans-3-Amino-cyclobutanecarboxylate hydrochloride 97%
Methyl trans-3-Amino-cyclobutanecarboxylate hydrochloride

Precaución: For research use only.

Formula: C₆H₁₁NO₂·ClH
MDL Number: MFCD17977174
CAS Number: 74316-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR