Benzyl 3-oxohexahydro-1H-pyrrolizine-7a-carboxylate 97%

Supplier: Ambeed
Warning

A1475236-100MG A1475236-250MG
76663-290EA 89.12 USD
76663-290 76663-292
Benzyl 3-oxohexahydro-1H-pyrrolizine-7a-carboxylate 97%
Benzyl 3-oxohexahydro-1H-pyrrolizine-7a-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₃
MW: 259.3 g/mol
MDL Number: MFCD11042349
CAS Number: 216392-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR