2,2-Difluoro-5-oxohexahydro-1H-pyrrolizine-7a-carboxylic acid 97%

Supplier: Ambeed
Warning

A1475230-100MG A1475230-1G A1475230-250MG
76663-284EA 53.05 USD
76663-284 76663-286 76663-288
2,2-Difluoro-5-oxohexahydro-1H-pyrrolizine-7a-carboxylic acid 97%
2,2-Difluoro-5-oxohexahydro-1H-pyrrolizine-7a-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉F₂NO₃
MW: 205.1588 g/mol
CAS Number: 2757082-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR