7a-(Hydroxymethyl)-2,2-dimethyltetrahydro-1H-pyrrolizin-3(2H)-one 97%
Supplier: Ambeed
|
Warning
|
A1475213-100MG
A1475213-250MG
76663-278EA
81.69
USD
76663-278
76663-280
7a-(Hydroxymethyl)-2,2-dimethyltetrahydro-1H-pyrrolizin-3(2H)-one 97%
7a-(Hydroxymethyl)-2,2-dimethyltetrahydro-1H-pyrrolizin-3(2H)-one
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₂ MW: 183.2475 g/mol |
CAS Number:
1788873-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...