2-Chloro-6-fluorophenylacetonitrile 98%

Supplier: Ambeed
Warning

A1475147-25G A1475147-5G
76663-274EA 43.5 USD
76663-274 76663-276
2-Chloro-6-fluorophenylacetonitrile 98%
2-Chloro-6-fluorophenylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MW: 169.59 g/mol
MDL Number: MFCD00001900
CAS Number: 75279-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR