1-Indanone 99%

Supplier: Ambeed
Warning

A1473982-100G A1473982-25G
76663-242EA 26 USD
76663-242 76663-244 76663-246
1-Indanone 99%
1-Indanone

Precaución: for Research Use Only

Formula: C₉H₈O
MDL Number: MFCD00003785
CAS Number: 83-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR