1-Bromo-2-cyclopropyl-4-fluoro-5-methoxybenzene 95%

Supplier: Ambeed
Warning

A1473921-1G A1473921-100MG A1473921-250MG
76663-234EA 1107 USD
76663-234 77875-112 77875-113
1-Bromo-2-cyclopropyl-4-fluoro-5-methoxybenzene 95%
1-Bromo-2-cyclopropyl-4-fluoro-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO
CAS Number: 1784088-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR