DL-Homoserine 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-HoSer-OH, H-DL-Hser-OH, H-DL-HomoSer-OH, (±)-Homoserine

A1473867-1G A1473867-25G A1473867-5G A1473867-10G
76663-228EA 6 USD
76663-228 76663-230 76663-232 77875-111
DL-Homoserine 98%
DL-Homoserine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Boiling Pt: 276
Melting Pt: 151
MDL Number: MFCD00002618
CAS Number: 1927-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR