(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Tyr(tBu)-OH, N-Boc-O-tert-butyl-L-tyrosine
A1473865-100G
A1473865-25G
A1473865-5G
76663-220EA
135
USD
76663-220
76663-222
76663-224
76663-226
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 98+%
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₈H₂₇NO₅ MW: 337.4 g/mol |
MDL Number:
MFCD00065598 CAS Number: 47375-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...