(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate 98%

Supplier: Ambeed
Warning

A1473863-100G A1473863-1G A1473863-25G A1473863-5G A1473863-500G
76663-212EA 119 USD
76663-212 76663-214 76663-216 76663-218 77875-110
(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate 98%
(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₅
MDL Number: MFCD03094770
CAS Number: 91229-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR