N-(tert-Butoxycarbonyl)-L-serine-β-lactone 98%

Supplier: Ambeed
Warning

Synonyms: (S)-ɑ-tert-Butoxycarbonylamino-β-propiolactone, tert-Butyl (S)-(2-oxooxetan-3-yl)carbamate, N-Boc-L-serineβ-Lactone, (S)-ɑ-Boc-amino-β-propiolactone, (S)-3-(tert-Butoxycarbonylamino)-2-oxetanone

A1473861-1G A1473861-5G
76663-208EA 54.11 USD
76663-208 76663-210
N-(tert-Butoxycarbonyl)-L-serine-β-lactone 98%
N-(tert-Butoxycarbonyl)-L-serine-beta-lactone

Precaución: for Research Use Only

Formula: C₈H₁₃NO₄
MW: 187.20 g/mol
MDL Number: MFCD01318414
CAS Number: 98541-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR