Boc-D-homoserine lactone 98+%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Homoserine lactone

A1473859-1G A1473859-25G A1473859-5G
76663-202EA 59.42 USD
76663-202 76663-204 76663-206
Boc-D-homoserine lactone 98+%
Boc-D-homoserine lactone

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
MDL Number: MFCD09037376
CAS Number: 67198-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR