(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride 98+%
Supplier: Ambeed
|
Warning
|
A1473857-100G
A1473857-1G
A1473857-25G
A1473857-5G
76663-194EA
610.02
USD
76663-194
76663-196
76663-198
76663-200
(R)-(+)-ɑ-Amino-γ-butyrolactone hydrochloride 98+%
(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₇NO₂·HCl |
MDL Number:
MFCD00674071 CAS Number: 104347-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...