Ni-(S)-BPB-Gly 98+%

Supplier: Ambeed
Warning

A1473855-1G A1473855-25G A1473855-5G
76663-188EA 35.01 USD
76663-188 76663-190 76663-192
Ni-(S)-BPB-Gly 98+%
Ni-(S)-BPB-Gly

Precaución: for Research Use Only

Formula: C₂₇H₂₅N₃NiO₃
MW: 498.19 g/mol
MDL Number: MFCD12545921
CAS Number: 96293-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR