Fmoc-L-Dap(Z)-OH 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Dap(Z)-OH
A1473841-1G
A1473841-250MG
A1473841-5G
76663-156EA
35
USD
76663-156
76663-158
76663-160
Fmoc-L-Dap(Z)-OH 98+%
Fmoc-L-Dap(Z)-OH
Precaución: for Research Use Only
Formula:
C₂₆H₂₄N₂O₆ MW: 460.50 g/mol |
MDL Number:
MFCD00273467 CAS Number: 204316-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...