Diisopropyl phosphonate 98%

Supplier: Ambeed
Warning

Synonyms: Diisopropyl phosphite, Di-i-propylphosphite, Diisopropyl hydrogen phosphite

A1473833-100G A1473833-25G A1473833-500G
76663-144EA 40.32 USD
76663-144 76663-146 76663-148
Diisopropyl phosphonate 98%
Diisopropyl phosphonate

Precaución: for Research Use Only

Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
MDL Number: MFCD00117905
CAS Number: 1809-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR