Methyl 1-aminocyclohexanecarboxylate hydrochloride 96+%

Supplier: Ambeed
Warning

A1473829-100G A1473829-1G A1473829-25G A1473829-5G
76663-136EA 621 USD
76663-136 76663-138 76663-140 76663-142
Methyl 1-aminocyclohexanecarboxylate hydrochloride 96+%
Methyl 1-aminocyclohexanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD09264353
CAS Number: 37993-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR