Lactitol monohydrate (contains < 10% H₂O)

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4R,5R)-4-(((2S,3R,4S,5R,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol hydrate

A1473805-10G A1473805-25G A1473805-100G A1473805-5G
76663-130EA 32 USD
76663-130 76663-132 77622-232 77875-107
Lactitol monohydrate (contains < 10% H₂O)
Lactitol monohydrate

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₁₁·H₂O
MW: 362.33 g/mol (344.31 anhy)
MDL Number: MFCD00150767
CAS Number: 81025-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR